Assay Details
| Gene Symbol: GABRA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260932.1 | NP_001247861.1 | 793 | 132 | ||
| XM_015141354.2 | XP_014996840.1 | 1174 | 132 | |||
| XM_028849019.1 | XP_028704852.1 | 821 | 132 | |||
| XM_028849020.1 | XP_028704853.1 | 1071 | 132 | |||
| XM_028849021.1 | XP_028704854.1 | 829 | 132 | |||
| GenBank mRNA | JU320577.1 | - | 1171 | 132 | ||
| JU320578.1 | - | 793 | 132 | |||
| JV703245.1 | - | 1207 | 132 | |||
More Information
Set Membership: |
|

GABRA1
Genomic Map