Assay Details
| Gene Symbol: RPAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257890.1 | NP_001244819.1 | 441 | 141 | ||
| GenBank mRNA | JU336440.1 | - | 441 | 141 | ||
| JU493688.1 | - | 325 | 141 | |||
| JV723696.1 | - | 417 | 141 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RPAP2
Genomic Map