Assay Details
| Gene Symbol: TMED5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266841.2 | NP_001253770.1 | 412 | 90 | ||
| GenBank mRNA | JU321772.1 | - | 447 | 90 | ||
| JU471143.1 | - | 459 | 90 | |||
| JV045307.1 | - | 412 | 90 | |||
More Information
Set Membership: |
|

TMED5
Genomic Map