Assay Details
| Gene Symbol: RFC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266677.1 | NP_001253606.1 | 729 | 85 | ||
| XM_015132410.2 | XP_014987896.1 | 695 | 85 | |||
| XM_015132411.2 | XP_014987897.1 | 836 | 85 | |||
| GenBank mRNA | JU335783.1 | - | 572 | 85 | ||
| JU476664.1 | - | 729 | 85 | |||
More Information
Set Membership: |
|

RFC4
Genomic Map