Assay Details
| Gene Symbol: SLC7A14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257912.1 | NP_001244841.1 | 837 | 95 | ||
| XM_028843109.1 | XP_028698942.1 | 1523 | 95 | |||
| GenBank mRNA | JU321422.1 | - | 837 | 95 | ||
More Information
Set Membership: |
|

SLC7A14
Genomic Map