Assay Details
| Gene Symbol: SLC6A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032823.1 | NP_001027995.1 | 1448 | 71 | ||
| XM_015118835.2 | XP_014974321.1 | 1832 | 71 | |||
| XM_015118836.2 | XP_014974322.1 | 1797 | 71 | |||
| XM_028835447.1 | XP_028691280.1 | 1731 | 71 | |||
| GenBank mRNA | AF285761.1 | - | 1448 | 71 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SLC6A4
Genomic Map