Assay Details
| Gene Symbol: IFI16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261661.1 | NP_001248590.1 | 1228 | 134 | ||
| GenBank mRNA | JU323500.1 | - | 1228 | 134 | ||
| JU471830.1 | - | 1258 | 134 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

IFI16
Genomic Map