Assay Details
| Gene Symbol: HSD3B7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125988.2 | XP_014981474.2 | 819 | 105 | ||
| GenBank mRNA | JV072133.1 | - | 887 | 105 | ||
| JV636388.1 | - | 895 | 105 | |||
More Information
Set Membership: |
|

HSD3B7
Genomic Map