Assay Details
| Gene Symbol: ZC3H15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258198.1 | NP_001245127.1 | 692 | 93 | ||
| GenBank mRNA | JU320303.1 | - | 690 | 93 | ||
| JU471527.1 | - | 692 | 93 | |||
| JV044633.1 | - | 657 | 93 | |||
More Information
Set Membership: |
|

ZC3H15
Genomic Map