Assay Details
| Gene Symbol: FBXO44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260876.1 | NP_001247805.1 | 689 | 73 | ||
| XM_015151844.2 | XP_015007330.1 | 435 | 73 | |||
| XM_015151906.2 | XP_015007392.1 | 483 | 73 | |||
| XM_015151959.2 | XP_015007445.1 | 413 | 73 | |||
| XM_015152037.2 | XP_015007523.1 | 769 | 73 | |||
| XM_015152103.2 | XP_015007589.1 | 539 | 73 | |||
| GenBank mRNA | JU319733.1 | - | 689 | 73 | ||
| JU321262.1 | - | 689 | 73 | |||
| JU496490.1 | - | 421 | 73 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

FBXO44
Genomic Map