Assay Details
| Gene Symbol: SMARCB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257888.2 | NP_001244817.1 | 930 | 63 | ||
| XM_015149824.2 | XP_015005310.1 | 1034 | 63 | |||
| GenBank mRNA | JU336021.1 | - | 957 | 63 | ||
| JU336022.1 | - | 930 | 63 | |||
| JU476885.1 | - | 914 | 63 | |||
| JU476886.1 | - | 887 | 63 | |||
| JV047994.1 | - | 980 | 63 | |||
| JV047995.1 | - | 953 | 63 | |||
More Information
Set Membership: |
|

SMARCB1
Genomic Map