Assay Details
| Gene Symbol: TMEM59L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194651.1 | NP_001181580.1 | 504 | 60 | ||
| XM_028838532.1 | XP_028694365.1 | 1489 | 60 | |||
| GenBank mRNA | JU326622.1 | - | 286 | 60 | ||
| JV463623.1 | - | 408 | 60 | |||
| JV565345.1 | - | 409 | 60 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TMEM59L
Genomic Map