Assay Details
| Gene Symbol: VSTM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194390.2 | NP_001181319.1 | 653 | 70 | ||
| XM_028853465.1 | XP_028709298.1 | 671 | 70 | |||
| GenBank mRNA | JU326897.1 | - | 569 | 70 | ||
| JV053978.1 | - | 611 | 70 | |||
| JV644558.1 | - | 623 | 70 | |||
More Information
Set Membership: |
|

VSTM4
Genomic Map