Assay Details
| Gene Symbol: MED26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015123227.2 | XP_014978713.1 | 563 | 95 | ||
| XM_028840032.1 | XP_028695865.1 | 343 | 95 | |||
| GenBank mRNA | JU477904.1 | - | 226 | 95 | ||
More Information
Set Membership: |
|

MED26
Genomic Map