Assay Details
| Gene Symbol: PPM1M | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193781.2 | NP_001180710.1 | 839 | 72 | ||
| XM_015130547.2 | XP_014986033.1 | 1445 | 72 | |||
| XM_028843141.1 | XP_028698974.1 | 1033 | 72 | |||
| XM_028843142.1 | XP_028698975.1 | 751 | 72 | |||
| GenBank mRNA | JU338879.1 | - | 754 | 72 | ||
| JU480427.1 | - | 771 | 72 | |||
| JU480428.1 | - | 757 | 72 | |||
| JV642449.1 | - | 750 | 72 | |||
More Information
Set Membership: |
|

PPM1M
Genomic Map