Assay Details
Gene Symbol: TFE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194297.2 | NP_001181226.1 | 1429 | 60 | ||
XM_015127335.2 | XP_014982821.1 | 1359 | 60 | |||
GenBank mRNA | JU336129.1 | - | 1194 | 60 | ||
JV044657.1 | - | 1340 | 60 | |||
JV528528.1 | - | 1374 | 60 | |||
JV668481.1 | - | 1374 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
