Assay Details
| Gene Symbol: LRWD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266042.1 | NP_001252971.1 | 1154 | 59 | ||
| XM_015134541.2 | XP_014990027.2 | 1421 | 59 | |||
| GenBank mRNA | JU473171.1 | - | 1154 | 59 | ||
| JV668665.1 | - | 1163 | 59 | |||
More Information
Set Membership: |
|

LRWD1
Genomic Map