Assay Details
| Gene Symbol: PKNOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261289.1 | NP_001248218.1 | 1040 | 82 | ||
| XM_015132797.2 | XP_014988283.2 | 1139 | 82 | |||
| XM_028845032.1 | XP_028700865.1 | 1400 | 82 | |||
| XM_028845033.1 | XP_028700866.1 | 1010 | 82 | |||
| GenBank mRNA | JU320847.1 | - | 1061 | 82 | ||
| JU471909.1 | - | 1040 | 82 | |||
| JU471910.1 | - | 1040 | 82 | |||
| JU476018.1 | - | 1040 | 82 | |||
More Information
Set Membership: |
|

PKNOX1
Genomic Map