Assay Details
| Gene Symbol: GPR180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001083934.4 | XP_001083934.1 | 1212 | 100 | ||
| GenBank mRNA | JU327148.1 | - | 817 | 100 | ||
| JU327149.1 | - | 817 | 100 | |||
| JV708154.1 | - | 860 | 100 | |||
More Information
Set Membership: |
|

GPR180
Genomic Map