Assay Details
| Gene Symbol: ZNF330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266710.1 | NP_001253639.1 | 405 | 84 | ||
| XM_015139223.2 | XP_014994709.1 | 401 | 84 | |||
| XM_015139224.2 | XP_014994710.1 | 599 | 84 | |||
| XM_015139225.2 | XP_014994711.1 | 395 | 84 | |||
| XM_015139227.2 | XP_014994713.1 | 358 | 84 | |||
| XM_028848014.1 | XP_028703847.1 | 427 | 84 | |||
| GenBank mRNA | JU334029.1 | - | 483 | 84 | ||
| JU475204.1 | - | 517 | 84 | |||
| JV046778.1 | - | 405 | 84 | |||
More Information
Set Membership: |
|

ZNF330
Genomic Map