Assay Details
| Gene Symbol: BMPR1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266263.1 | NP_001253192.1 | 1535 | 76 | ||
| GenBank mRNA | JU336012.1 | - | 1360 | 76 | ||
| JV043223.1 | - | 1535 | 76 | |||
| JV638900.1 | - | 1574 | 76 | |||
More Information
Set Membership: |
|

BMPR1B
Genomic Map