Assay Details
| Gene Symbol: H2AFY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266414.1 | NP_001253343.1 | 814 | 87 | ||
| GenBank mRNA | JU335342.1 | - | 697 | 87 | ||
| JV047575.1 | - | 814 | 87 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

H2AFY2
Genomic Map