Assay Details
| Gene Symbol: ARF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015134850.2 | XP_014990336.1 | 308 | 114 | ||
| GenBank mRNA | JU320256.1 | - | 182 | 114 | ||
| JU470460.1 | - | 187 | 114 | |||
| JV044714.1 | - | 189 | 114 | |||
| JV566605.1 | - | 201 | 114 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ARF5
Genomic Map