Assay Details
| Gene Symbol: ATP1B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015118521.2 | XP_014974007.1 | 1028 | 81 | ||
| GenBank mRNA | JU321993.1 | - | 923 | 81 | ||
| JV508044.1 | - | 925 | 81 | |||
| JV567491.1 | - | 926 | 81 | |||
More Information
Set Membership: |
|

ATP1B2
Genomic Map