Assay Details
| Gene Symbol: OTUB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001095170.4 | XP_001095170.1 | 503 | 66 | ||
| GenBank mRNA | JV661106.1 | - | 292 | 66 | ||
| JV704617.1 | - | 318 | 66 | |||
More Information
Set Membership: |
|

OTUB2
Genomic Map