Assay Details
| Gene Symbol: SULT4A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015150530.2 | XP_015006016.2 | 738 | 73 | ||
| GenBank mRNA | JU320831.1 | - | 647 | 73 | ||
| JV567444.1 | - | 673 | 73 | |||
More Information
Set Membership: |
|

SULT4A1
Genomic Map