Assay Details
Gene Symbol: C1QTNF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015139816.2 | XP_014995302.1 | 1114 | 90 | ||
XM_015139817.2 | XP_014995303.2 | 295 | 90 | |||
XM_015139819.2 | XP_014995305.1 | 237 | 90 | |||
XM_028849368.1 | XP_028705201.1 | 220 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
