Assay Details
| Gene Symbol: AP2M1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260598.1 | NP_001247527.1 | 1219 | 65 | ||
| XM_015131437.2 | XP_014986923.1 | 1388 | 65 | |||
| GenBank mRNA | JU332849.1 | - | 1220 | 65 | ||
| JU332850.1 | - | 1214 | 65 | |||
| JU474249.1 | - | 1228 | 65 | |||
| JU474250.1 | - | 1222 | 65 | |||
| JV046138.1 | - | 1225 | 65 | |||
| JV046139.1 | - | 1219 | 65 | |||
| JV641775.1 | - | 1225 | 65 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

AP2M1
Genomic Map