Assay Details
| Gene Symbol: PELI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257883.2 | NP_001244812.1 | 501 | 101 | ||
| XM_015113537.2 | XP_014969023.1 | 630 | 101 | |||
| GenBank mRNA | JU333768.1 | - | 503 | 101 | ||
| JV565038.1 | - | 586 | 101 | |||
More Information
Set Membership: |
|

PELI3
Genomic Map