Assay Details
| Gene Symbol: STARD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001115049.4 | XP_001115049.1 | 956 | 71 | ||
| XM_015115153.2 | XP_014970639.1 | 2150 | 71 | |||
| XM_015115157.2 | XP_014970643.1 | 896 | 71 | |||
| XM_015115160.2 | XP_014970646.1 | 833 | 71 | |||
| GenBank mRNA | JU321576.1 | - | 1529 | 71 | ||
| JU474077.1 | - | 741 | 71 | |||
| JV045124.1 | - | 823 | 71 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

STARD10
Genomic Map