Assay Details
| Gene Symbol: B4GALT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015137763.2 | XP_014993249.1 | 671 | 65 | ||
| XM_028834533.1 | XP_028690366.1 | 467 | 65 | |||
| XM_028834538.1 | XP_028690371.1 | 467 | 65 | |||
| XM_028834540.1 | XP_028690373.1 | 660 | 65 | |||
| GenBank mRNA | JV524214.1 | - | 324 | 65 | ||
| JV632097.1 | - | 531 | 65 | |||
More Information
Set Membership: |
|

B4GALT2
Genomic Map