Assay Details
| Gene Symbol: RNF128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135114.1 | NP_001128586.1 | 821 | 81 | ||
| XM_015127920.2 | XP_014983406.1 | 996 | 81 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RNF128
Genomic Map