Assay Details
| Gene Symbol: ALG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257868.1 | NP_001244797.1 | 582 | 91 | ||
| XM_015120883.2 | XP_014976369.1 | 558 | 91 | |||
| GenBank mRNA | JU322609.1 | - | 582 | 91 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ALG5
Genomic Map