Assay Details
| Gene Symbol: BAHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257462.2 | NP_001244391.1 | 1990 | 62 | ||
| XM_015141871.2 | XP_014997357.1 | 2355 | 62 | |||
| XM_015141872.2 | XP_014997358.1 | 2223 | 62 | |||
| XM_015141873.2 | XP_014997359.1 | 2366 | 62 | |||
| XM_015141874.2 | XP_014997360.1 | 2274 | 62 | |||
| XM_015141877.2 | XP_014997363.1 | 2231 | 62 | |||
| XM_028850301.1 | XP_028706134.1 | 2014 | 62 | |||
| XM_028850302.1 | XP_028706135.1 | 2011 | 62 | |||
| XM_028850303.1 | XP_028706136.1 | 1989 | 62 | |||
| XM_028850304.1 | XP_028706137.1 | 2124 | 62 | |||
| GenBank mRNA | JU321091.1 | - | 1990 | 62 | ||
| JU333471.1 | - | 1990 | 62 | |||
| JU471184.1 | - | 1993 | 62 | |||
| JU474747.1 | - | 1993 | 62 | |||
More Information
Set Membership: |
|

BAHD1
Genomic Map