Assay Details
| Gene Symbol: P3H4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028836480.1 | XP_028692313.1 | 1512 | 61 | ||
| XM_028836481.1 | XP_028692314.1 | 932 | 61 | |||
| GenBank mRNA | JU326395.1 | - | 239 | 61 | ||
| JV045541.1 | - | 467 | 61 | |||
| JV454551.1 | - | 44 | 61 | |||
| JV569158.1 | - | 76 | 61 | |||
More Information
Set Membership: |
|

P3H4
Genomic Map