Assay Details
| Gene Symbol: MTFP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015150151.2 | XP_015005637.2 | 488 | 65 | ||
| XM_015150152.2 | XP_015005638.1 | 488 | 65 | |||
| GenBank mRNA | JU475926.1 | - | 122 | 65 | ||
| JV045362.1 | - | 148 | 65 | |||
More Information
Set Membership: |
|

MTFP1
Genomic Map