Assay Details
| Gene Symbol: ANKH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261260.1 | NP_001248189.1 | 604 | 68 | ||
| GenBank mRNA | JU320371.1 | - | 627 | 68 | ||
| JU471381.1 | - | 604 | 68 | |||
More Information
Set Membership: |
|

ANKH
Genomic Map