Assay Details
| Gene Symbol: LCA5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194409.2 | NP_001181338.1 | 1588 | 153 | ||
| XM_015132855.2 | XP_014988341.1 | 1722 | 153 | |||
| XM_015132856.2 | XP_014988342.1 | 1521 | 153 | |||
| XM_015132864.2 | XP_014988350.1 | 2060 | 153 | |||
| XM_015132866.2 | XP_014988352.1 | 1912 | 153 | |||
| XM_028845049.1 | XP_028700882.1 | 1684 | 153 | |||
| XM_028845050.1 | XP_028700883.1 | 1888 | 153 | |||
| XM_028845051.1 | XP_028700884.1 | 1725 | 153 | |||
| XM_028845052.1 | XP_028700885.1 | 1425 | 153 | |||
| XM_028845053.1 | XP_028700886.1 | 1690 | 153 | |||
| XM_028845054.1 | XP_028700887.1 | 1490 | 153 | |||
| XM_028845055.1 | XP_028700888.1 | 1535 | 153 | |||
| XM_028845056.1 | XP_028700889.1 | 1348 | 153 | |||
| XM_028845057.1 | XP_028700890.1 | 1201 | 153 | |||
| XM_028845058.1 | XP_028700891.1 | 338 | 153 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028845058.1 transcripts.
Set Membership: |
|

LCA5L
Genomic Map