Assay Details
| Gene Symbol: CDKN2AIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261581.1 | NP_001248510.1 | 563 | 103 | ||
| GenBank mRNA | JU322461.1 | - | 557 | 103 | ||
| JU470704.1 | - | 563 | 103 | |||
More Information
Set Membership: |
|

CDKN2AIP
Genomic Map