Assay Details
| Gene Symbol: SMAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261367.1 | NP_001248296.1 | 1285 | 99 | ||
| XM_015139264.2 | XP_014994750.1 | 1750 | 99 | |||
| XM_015139265.2 | XP_014994751.1 | 1806 | 99 | |||
| XM_015139266.2 | XP_014994752.1 | 3624 | 99 | |||
| XM_015139267.2 | XP_014994753.1 | 2299 | 99 | |||
| GenBank mRNA | AY369827.1 | - | 562 | 99 | ||
| JU319940.1 | - | 1447 | 99 | |||
| JU475606.1 | - | 1439 | 99 | |||
| JV047058.1 | - | 1285 | 99 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SMAD1
Genomic Map