Assay Details
| Gene Symbol: TP53BP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001093747.4 | XP_001093747.2 | 3140 | 84 | ||
| XM_015115521.2 | XP_014971007.1 | 2992 | 84 | |||
| GenBank mRNA | JU321062.1 | - | 2921 | 84 | ||
| JU470516.1 | - | 2965 | 84 | |||
| JV044010.1 | - | 2914 | 84 | |||
| JV627428.1 | - | 2091 | 84 | |||
| JV660194.1 | - | 3138 | 84 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TP53BP2
Genomic Map