Assay Details
| Gene Symbol: KLF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265918.2 | NP_001252847.1 | 574 | 92 | ||
| GenBank mRNA | JU336104.1 | - | 638 | 92 | ||
| JU473578.1 | - | 591 | 92 | |||
| JV045548.1 | - | 574 | 92 | |||
| JV638150.1 | - | 839 | 92 | |||
More Information
Set Membership: |
|

KLF9
Genomic Map