Assay Details
| Gene Symbol: CCT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015112024.2 | XP_014967510.1 | 1428 | 80 | ||
| GenBank mRNA | JU321928.1 | - | 1219 | 80 | ||
| JU470795.1 | - | 1270 | 80 | |||
| JV045035.1 | - | 1221 | 80 | |||
| JV602218.1 | - | 1244 | 80 | |||
| JV638590.1 | - | 1313 | 80 | |||
More Information
Set Membership: |
|

CCT3
Genomic Map