Assay Details
| Gene Symbol: AMACR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266997.1 | NP_001253926.1 | 807 | 97 | ||
| XM_015139802.2 | XP_014995288.1 | 656 | 97 | |||
| XM_028849128.1 | XP_028704961.1 | 824 | 97 | |||
| XM_028849129.1 | XP_028704962.1 | 650 | 97 | |||
| GenBank mRNA | JU323444.1 | - | 646 | 97 | ||
| JU323472.1 | - | 807 | 97 | |||
| JV704471.1 | - | 649 | 97 | |||
More Information
Set Membership: |
|

AMACR
Genomic Map