Assay Details
| Gene Symbol: UBA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265961.1 | NP_001252890.1 | 718 | 69 | ||
| XM_015130400.2 | XP_014985886.1 | 693 | 69 | |||
| XM_015130401.2 | XP_014985887.1 | 669 | 69 | |||
| XM_015130402.2 | XP_014985888.1 | 613 | 69 | |||
| GenBank mRNA | JU328580.1 | - | 634 | 69 | ||
| JU470540.1 | - | 718 | 69 | |||
More Information
Set Membership: |
|

UBA3
Genomic Map