Assay Details
| Gene Symbol: MAN1A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261288.1 | NP_001248217.1 | 1331 | 131 | ||
| XM_028844502.1 | XP_028700335.1 | 2104 | 131 | |||
| GenBank mRNA | JU332633.1 | - | 1345 | 131 | ||
| JU472018.1 | - | 1364 | 131 | |||
| JV045346.1 | - | 1331 | 131 | |||
More Information
Set Membership: |
|

MAN1A2
Genomic Map