Assay Details
| Gene Symbol: PUS7L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015151515.2 | XP_015007001.2 | 2205 | 145 | ||
| XM_015151516.2 | XP_015007002.2 | 2205 | 145 | |||
| XM_015151517.2 | XP_015007003.2 | 2205 | 145 | |||
| GenBank mRNA | JV724586.1 | - | 1108 | 145 | ||
More Information
Set Membership: |
|

PUS7L
Genomic Map