Assay Details
| Gene Symbol: CDH22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015148909.2 | XP_015004395.1 | 1868 | 55 | ||
| XM_015148910.2 | XP_015004396.1 | 1870 | 55 | |||
| XM_015148911.2 | XP_015004397.1 | 5532 | 55 | |||
| XM_028828456.1 | XP_028684289.1 | 1287 | 55 | |||
| GenBank mRNA | JV625741.1 | - | 1435 | 55 | ||
More Information
Set Membership: |
|

CDH22
Genomic Map