Assay Details
| Gene Symbol: OTULINL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193791.3 | NP_001180720.1 | 356 | 97 | ||
| XM_015139746.2 | XP_014995232.1 | 356 | 97 | |||
| GenBank mRNA | JU326128.1 | - | 227 | 97 | ||
| JV063818.1 | - | 250 | 97 | |||
| JV654192.1 | - | 217 | 97 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

OTULINL
Genomic Map