Assay Details
| Gene Symbol: CIB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257882.1 | NP_001244811.1 | 624 | 74 | ||
| XM_015142817.2 | XP_014998303.1 | 662 | 74 | |||
| XM_015142818.2 | XP_014998304.1 | 537 | 74 | |||
| GenBank mRNA | JU332526.1 | - | 624 | 74 | ||
| JU474000.1 | - | 612 | 74 | |||
More Information
Set Membership: |
|

CIB2
Genomic Map